Learn More
Greiner Bio-One Zip Bags
Product Code. 12529146
Change view
Click to view available options
Dimensions (L x W):
120 x 180mm
Unit Size:
Pack of 1000
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Unit Size
12529146 120 x 180mm Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12529146 Supplier Greiner Bio-One Supplier No. SSPA120
  • ONLINE EXCLUSIVE PRICE
    234.15€
    129.80€ / Pack of 1000
    Save 104.35€
    44% Off
Estimated Shipment: 21-09-2026
Specifications
Closure Type Zip Seal
Quantity 1000 Pk.
Dimensions (L x W) 120 x 180mm
Product Type Zip-seal Storage Bag