missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZMPSTE24 Antibody, mFluor Violet 610 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-2388MFV610
This item is not returnable.
View return policy
Description
ZMPSTE24 Polyclonal antibody specifically detects ZMPSTE24 in Human samples. It is validated for Western Blot
Specifications
| ZMPSTE24 | |
| Polyclonal | |
| Western Blot | |
| zona pellucida binding protein 2, zona pellucida-binding protein 2, ZPBP-like protein, ZPBPLMGC41930 | |
| A synthetic peptide made to a C-terminal portion of the human ZMPSTE24 protein sequence (between residues 400-475). [Swiss-Prot# O75844] | |
| 0.1 mL | |
| Angiogenesis, Cancer, Cell Biology, Cellular Markers, Extracellular Matrix, Signal Transduction, Ubiquitin Proteasome Pathway | |
| 10269 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction