missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF10 Antibody (CL9747) [Janelia Fluor™ 525], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-88940JF525
This item is not returnable.
View return policy
Description
ZNF10 Monoclonal antibody specifically detects ZNF10 in Human samples. It is validated for Western Blot,Immunocytochemistry/ Immunofluorescence
Specifications
| ZNF10 | |
| Monoclonal | |
| Janelia Fluor 525 | |
| KOX1zinc finger protein 10 (KOX 1), zinc finger protein 10, Zinc finger protein KOX1 | |
| This antibody was developed using a synthetic peptide derived from P21506, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Transcription Factors and Regulators | |
| 7556 | |
| Store at 4⪚C in the dark. | |
| IgG2a |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| CL9747 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction