missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF124 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25216-1-AP-150UL
This item is not returnable.
View return policy
Description
May be involved in transcriptional regulation.Specifications
| ZNF124 | |
| Polyclonal | |
| Unconjugated | |
| ZNF124 | |
| HZF 16, HZF16, zinc finger protein 124, Zinc finger protein HZF 16, ZNF124 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 7678 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.26 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q15973 | |
| ZNF124 | |
| ZNF124 Fusion Protein Ag17165 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction