Learn More
ZNF18 Monoclonal Antibody (OTI4C5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA810108
Description
ZNF18 be involved in transcriptional regulation.Specifications
| ZNF18 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P17022 | |
| ZNF18 | |
| Human recombit protein fragment corresponding to amino acids 183-405 of human ZNF18 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI4C5 | |
| Unconjugated | |
| ZNF18 | |
| HDSG1, KOX11, ZKSCAN6, ZNF535, ZSCAN38, Zfp535 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 7566 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.