missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZNF184 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA584670
This item is not returnable.
View return policy
Description
Immunogen sequence: MEDLSSPDST LLQGGHNLLS SASFQ Highest antigen sequence indentity to the following orthologs: Mouse - 44%, Rat - 44%.
The protein encoded by this gene is predicted to be a Kruppel C2H2-type zinc-finger protein family member. Sequence analysis predicts that the protein contains two Kruppel associated box (KRAB) boxes in the N-terminus and highly conserved zinc finger motifs at the C-terminus. Alternative splicing results in multiple transcript variants.
Specifications
| ZNF184 | |
| Polyclonal | |
| Unconjugated | |
| ZNF184 | |
| kr-ZNF3; Zinc finger protein 184; zinc finger protein 184 (Kruppel-like); ZNF184 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 7738 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ChIP Assay, Immunocytochemistry | |
| 0.4 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q99676 | |
| ZNF184 | |
| Recombinant protein corresponding to Human ZNF184. Recombinant protein control fragment (Product #RP-108503). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction