missing translation for 'onlineSavingsMsg'
Learn More

ZNF23 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

145.00€ - 410.00€

Specifications

Antigen ZNF23
Concentration 0.32 mg/mL
Applications Western Blot, Immunocytochemistry, Immunoprecipitation, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16870925
View Documents
Proteintech
21322-1-AP-20UL
20 μL
145.00€
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16860925
View Documents
Proteintech
21322-1-AP-150UL
150 μL
410.00€
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

ZNF23 May be involved in transcriptional regulation. May have a role in embryonic development. Belongs to the krueppel C2H2-type zinc-finger protein family.
Specifications

Specifications

ZNF23
Western Blot, Immunocytochemistry, Immunoprecipitation, Immunofluorescence
Unconjugated
Rabbit
Mouse, Rat, Human
P17027
234725, 307839, 7571
ZNF23 Fusion Protein Ag14755
Primary
-20°C
ZNF23
0.32 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
KOX16, Zfp612, Zinc finger protein 23, Zinc finger protein 359, Zinc finger protein KOX16, ZNF23, ZNF359, ZNF612
Zfp612, ZNF23
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.