missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF259 Antibody (LG1) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-22448-0.025mg
This item is not returnable.
View return policy
Description
ZNF259 Monoclonal specifically detects ZNF259 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| ZNF259 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500 - 1:2000, Immunocytochemistry/Immunofluorescence 1:500 | |
| ERP1, EZFIT-related protein 1, KIAA1396MGC150657, MGC150658, Z1971, zinc finger protein 471 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 8882 | |
| Human | |
| Purified |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| LG1 | |
| Unconjugated | |
| O75312 | |
| ZNF259 | |
| murine ZPR1 protein | |
| 0.025 mg | |
| Primary | |
| This antibody reacts with Human. | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction