missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF324 Antibody, FITC, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-78183F
This item is not returnable.
View return policy
Description
ZNF324 Polyclonal antibody specifically detects ZNF324 in Human samples. It is validated for Western Blot, Immunoprecipitation (Negative)
Specifications
| ZNF324 | |
| Polyclonal | |
| Western Blot, Immunoprecipitation (Negative) | |
| ZF5128, zinc finger protein 324, zinc finger protein 324A, zinc finger protein ZF5128, ZNF324A | |
| The immunogen for this product maps to a region between residue 503 and 553 of human Zinc Finger Protein 324 using the numbering given in entry NP_055162.1 (GeneID 25799). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| FITC | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 25799 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction