Learn More
ZNF354A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 14072-1-AP-150UL
 View more versions of this product
Product Code. 16802554
  • 410.00€ / 150µL
Description
ZNF354A gene ontology annotations related to this gene include regulation of transcription by RNA polymerase II; sensory perception of sound.
Specifications
Specifications
ZNF354A
Polyclonal
Unconjugated
ZNF354A
EZNF, HKL1, KID 1, KID1, TCF 17, TCF17, Transcription factor 17, zinc finger protein 354A, Zinc finger protein eZNF, ZNF354A
Rabbit
Antigen Affinity Chromatography
RUO
21408, 24522, 6940
-20°C
Liquid
Western Blot
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O60765, Q02975, Q61751
ZNF354A
ZNF354A Fusion Protein Ag5142
150 μL