Learn More
ZNF385B Monoclonal Antibody (OTI2G4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA802727S
Description
ZNF385B Monoclonal antibody specifically detects ZNF385B in Human samples. It is validated for Western BlotSpecifications
| ZNF385B | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q569K4 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 151126 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI2G4 | |
| Unconjugated | |
| ZNF385B | |
| B830010L13Rik; C130013B13Rik; si:busm1-42h5.1; si:by51i9.1; si:dz145b16.1; si:dz42h5.1; si:xx-51i9.1; wu:fj47d04; ZFN533; Zfp385b; Zfp533; zgc:73248; Zinc finger protein 385B; zinc finger protein 533; ZNF385B; ZNF533 | |
| Human recombinant protein fragment corresponding to amino acids 149-399 of human ZNF385B produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.