missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF420 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
280.00€ - 624.00€
Specifications
| Antigen | ZNF420 |
|---|---|
| Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18238022
|
Novus Biologicals
NBP2-55984 |
100 μL |
624.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18676216
|
Novus Biologicals
NBP2-55984-25ul |
25 μL |
280.00€
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ZNF420 Polyclonal specifically detects ZNF420 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| ZNF420 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| FLJ20086, KIAA1584, MGC21637, MGC61687, SUHW4, Suppressor of hairy wing homolog 4, zinc finger protein 280D, Zinc finger protein 634, ZNF634suppressor of hairy wing homolog 4 (Drosophila) | |
| ZNF420 | |
| IgG | |
| Affinity Purified |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| 147923 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:TAFLAGQQDRSDIYMSRRFTKSIFPVFFSVPTMWTSKLELRYSHGDFENGIWVLWIMEQKYRRSLQP | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title