missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF443 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13667-1-AP-150UL
This item is not returnable.
View return policy
Description
Zinc finger proteins (ZNFs) bind DNA and, through this binding, regulate gene transcription. Most ZNFs contain conserved C2H2 motifs and are classified as Kruppel-type zinc fingers. For a general description of these proteins, see ZNF91.Specifications
| ZNF443 | |
| Polyclonal | |
| Unconjugated | |
| ZNF443 | |
| zinc finger protein 443, ZK1, ZNF443 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10224 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9Y2A4 | |
| ZNF443 | |
| ZNF443 Fusion Protein Ag4585 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction