Learn More
ZNF480 Monoclonal Antibody (OTI7H4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA812669S
Description
ZNF480 Monoclonal antibody specifically detects ZNF480 in Human samples. It is validated for Western BlotSpecifications
| ZNF480 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q8WV37 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 147657 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2a |
| Western Blot | |
| OTI7H4 | |
| Unconjugated | |
| ZNF480 | |
| zinc finger protein 480; ZNF480 | |
| Full length human recombinant protein of human ZNF480 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.