Learn More
ZNF639 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA562564
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: KRKTLHPSRY SDSSGISRIA DGFNGIFSDH CYSVCSMRQP DLKYFDNKDD DSDTETSNDL PKFADGIKAR NRNQNYLVPS PVL Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 88%.
This gene encodes a multi-domain scaffolding protein that is required for normal development. This protein may have a role in septate junction formation, signal transduction, cell proliferation, synaptogenesis and lymphocyte activation. Several alternatively spliced transcript variants encoding different isoforms have been described for this gene, but the full-length nature of some of the variants is not known.Specifications
ZNF639 | |
Polyclonal | |
Unconjugated | |
ZNF639 | |
6230400O18Rik; ANC_2H01; ANC-2H01; ZASC1; Zfp639; zinc finger amplified in esophageal squamous cell carcinomas 1; zinc finger protein 639; Zinc finger protein 639-like protein; Zinc finger protein ANC_2H01; Zinc finger protein ZASC1; ZNF639 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
51193 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9UID6 | |
ZNF639 | |
Recombinant protein corresponding to Human ZNF639 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only