Learn More
ZNF641 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26302-1-AP-150UL
Description
The ZNF641 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human ZNF641. This antibody recognizes human, mouse , rat antigen. The ZNF641 antibody has been validated for the following applications: IF, WB, ELISA analysis.
Specifications
| ZNF641 | |
| 800 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Mouse, Rat | |
| 150 kDa | |
| IgG |
| Western Blot, Immunofluorescence, ELISA | |
| zinc finger protein 641, ZNF641 | |
| 150 μL | |
| 121274 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |