missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF655 Antibody, FITC, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-78732F
This item is not returnable.
View return policy
Description
ZNF655 Polyclonal antibody specifically detects ZNF655 in Human samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| ZNF655 | |
| Polyclonal | |
| Western Blot, Immunoprecipitation | |
| zinc finger protein 655 | |
| The immunogen for this product maps to a region between residue 1 and 50 of human Vav-1 Interacting Kruppel-like Protein using the numbering given in entry NP_612503.1 (GeneID 79027). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunoprecipitation | |
| FITC | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 79027 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction