missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF846 Mouse anti-Human, Clone: ZNF846/2687, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-07853-100ug
This item is not returnable.
View return policy
Description
ZNF846 Monoclonal specifically detects ZNF846 in Human samples. It is validated for Immunohistochemistry-Paraffin, Protein Array.
Specifications
| ZNF846 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Immunohistochemistry-Paraffin 1 to 2 μg/mL, Protein Array | |
| zinc finger protein 846 | |
| Recombinant full-length human ZNF846 protein | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin), Peptide Array | |
| ZNF846/2687 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 162993 | |
| Store at 4°C. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction