missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNFN1A4 Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP3-12160
This item is not returnable.
View return policy
Description
ZNFN1A4 Polyclonal antibody specifically detects ZNFN1A4 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Peptide ELISA
Specifications
| ZNFN1A4 | |
| Polyclonal | |
| Western Blot 0.1 - 2 μg/mL, Flow Cytometry 10 μg/mL, Immunocytochemistry/ Immunofluorescence 5 μg/mL, Peptide ELISA Detection limit 1:2000 | |
| EOS, IKAROS family zinc finger 4 (Eos), Ikaros family zinc finger protein 4, KIAA1782Eos, zinc finger protein, subfamily 1A, 4 (Eos), zinc finger transcription factor Eos, ZNFN1A4zinc finger protein Eos | |
| Peptide with sequence C-RPTFIDRLANSLTKR corresponding to internal region according to NP_071910.3. | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, Flow Cytometry, Immunofluorescence, Peptide ELISA | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA | |
| Goat | |
| Immunogen affinity purified | |
| RUO | |
| 64375 | |
| Store at -20°C in the dark. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction