missing translation for 'onlineSavingsMsg'
Learn More

Zoalene (500 ug/mL in Methanol), TRC

Product Code. 30398222
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30398222 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30398222 Supplier Toronto Research Chemicals Supplier No. TRCKIT09581X1ML
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 148-01-6
Molecular Formula C8 H7 N3 O5
Quantity 1 mL
Synonym Benzamide, 2-methyl-3,5-dinitro-, o-Toluamide, 3,5-dinitro- (6CI,8CI), 2-Methyl-3,5-dinitrobenzamide, 3,5-Dinitro-2-methylbenzamide, 3,5-Dinitro-o-toluamide, Coccidin, Coccidine A, Coccidot, Dinitolmide, Salcostat, Zoalene, Zoamix
InChI Formula InChI=1S/C8H7N3O5/c1-4-6(8(9)12)2-5(10(13)14)3-7(4)11(15)16/h2-3H,1H3,(H2,9,12)
SMILES Cc1c(cc(cc1[N+](=O)[O-])[N+](=O)[O-])C(=O)N
IUPAC Name 2-methyl-3,5-dinitrobenzamide
Molecular Weight (g/mol) 225.16
Formula Weight 225.0386
Concentration or Composition (by Analyte or Components) 3,5-Dinitro-o-toluamide 500 μg/mL
Chemical Name or Material 3,5-Dinitro-o-toluamide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.