missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Zoledronic Acid
GREENER_CHOICE

Product Code. 18720109
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18720109 50 mg 50mg
18711368 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27950” to get your promotional price
Product Code. 18720109 Supplier Tocris Bioscience™ Supplier No. 6111/50
  • ONLINE EXCLUSIVE PRICE
    410.00€
    387.45€ / 50mg
    Save 22.55€
    5% Off
View all promotions
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27950” to get your promotional price

Potent farnesyl diphosphate (FPP) synthase inhibitor

Zoledronic Acid is a potent bisphophonate farnesyl diphosphate (FPP) synthase inhibitor (IC50 = 20 nM). Inhibits osteoclast-mediated bone resorption. Also inhibits Ras signaling and tumor growth, and induces apoptosis in pancreatic cancer cells. Reverses epithelial-mesenchymal transition and inhibits breast cancer cell renewal via inactivation of NF-κB.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ras GTPase Inhibitors
CAS 118072-93-8
Synonym [1-Hydroxy-2-(1H-imidazol-1-yl)ethylidene]bisphosphonic acid
Quantity 50 mg
Solubility Soluble to 50mM in 1.1eq. NaOH and to 3mM in water
Molecular Formula C5H10N2O7P2
Formula Weight 272.09
Physical Form Solid
Color Off-white
Product Type Bioactive Small Molecule
Purity >98%
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.