missing translation for 'onlineSavingsMsg'
Learn More

Zoledronic acid, TRC

Product Code. 30137954
Change view
Click to view available options
Quantity:
2.5 mg
5 mg
25 mg
Unit Size:
2.5mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137954 25 mg 25mg
30012705 2.5 mg 2.5mg
30009595 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137954 Supplier Toronto Research Chemicals Supplier No. TRCZ62350525MG

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 118072-93-8
Molecular Formula C5 H10 N2 O7 P2
Molecular Weight (g/mol) 272.09
Synonym Phosphonic acid, [1-hydroxy-2-(1H-imidazol-1-yl)ethylidene]bis- (9CI), (1-Hydroxy-2-imidazol-1-ylethylidene)diphosphonic acid, 1-Hydroxy-2-(1H-imidazol-1-yl)ethylidene-1,1-bisphosphonic acid, 2-(Imidazol-1-yl)-1-hydroxy-1,1'-ethylidenediphosphonic acid, Aclasta, BPH 91, CGP 42446, Orazol, Reclast, ZOL 446, Zoledronate, Zoledronic acid, Zometa, Phosphonic acid, P,P'-[1-hydroxy-2-(1H-imidazol-1-yl)ethylidene]bis-
IUPAC Name (1-hydroxy-2-imidazol-1-yl-1-phosphonoethyl)phosphonic acid
SMILES OC(P(O)(O)=O)(P(O)(O)=O)CN1C=CN=C1

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C5H10N2O7P2/c8-5(15(9,10)11,16(12,13)14)3-7-2-1-6-4-7/h1-2,4,8H,3H2,(H2,9,10,11)(H2,12,13,14)
Formula Weight 271.9963
Chemical Name or Material Zoledronic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.