missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Zolmitriptan

Product Code. 15433359
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15433359 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15433359 Supplier Thermo Scientific Chemicals Supplier No. J60616.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A serotonin 5HT1D-receptor agonist | CAS: 139264-17-8 | C16H21N3O2

A serotonin 5HT1D-receptor agonist

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 g
Description White
For Use With (Application) A serotonin 5HT1D-receptor agonist
CAS 139264-17-8
Molecular Formula C16H21N3O2
Formula Weight 287.36g/mol
Synonym (S)-4-(3-[2-(Dimethylamino)ethyl]-5-indolylmethyl)oxazolidin-2-one
Melting Point 136°C to 141°C
Solubility Soluble in DMSO at 5mg/ml
Product Type Signal Transduction Reagent
Form Crystalline powder
Show More Show Less

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.