missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZRANB1/Trabid Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
415.00€ - 572.00€
Specifications
| Antigen | ZRANB1/Trabid |
|---|---|
| Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18444211
|
Novus Biologicals
NBP1-86918-25ul |
25 μL |
415.00€
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18275308
|
Novus Biologicals
NBP1-86918 |
0.1 mL |
572.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ZRANB1/Trabid Polyclonal specifically detects ZRANB1/Trabid in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| ZRANB1/Trabid | |
| Polyclonal | |
| Rabbit | |
| Cancer, DNA Repair, DNA replication Transcription Translation and Splicing | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 54764 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:SSGNSQRRSPPATKRDSEVKMDFQRIELAGAVGSKEELEVDFKKLKQIKNRMKKTDWLFLNACVGVVEGDLAAIEAYKSSGGDIARQLT | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Unconjugated | |
| RUO | |
| Human | |
| HRPE773, jacalin-like lectin domain containing 2, JCLN2, pancreatic adenocarcinoma upregulated factor, PAUF, PRO1567, zymogen granule protein 16 homolog B, zymogen granule protein 16 homolog B (rat) | |
| ZRANB1 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title