Learn More
Axygen™ Zymark™ Rapid Plate/Sciclone and Tecan™ TEMO Acquarius Genmate: 100 μL
Product Code. 12736769 Shop All Axygen Scientific Products
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
Tip Style:
Maxymum Recovery, Round
Standard, Round
Filtered:
Filtered
Non-filtered
Unit Size:
Pack of 4800
Pack of 960
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sterility Tip Style Filtered Unit Size
12736769 Sterile Standard, Round Filtered Pack of 4800
11354335 Sterile Maxymum Recovery, Round Filtered Pack of 960
12726769 Sterile Maxymum Recovery, Round Filtered Pack of 4800
11344325 Sterile Standard, Round Non-filtered Pack of 960
12786669 Non-sterile Standard, Round Non-filtered Pack of 4800
11334325 Non-sterile Standard, Round Non-filtered Pack of 960
12545577 Non-sterile Maxymum Recovery, Round Non-filtered Pack of 960
11346064 Non-sterile Maxymum Recovery, Round Non-filtered Pack of 4800
11364335 Sterile Standard, Round Filtered Pack of 960
11324325 Sterile Maxymum Recovery, Round Non-filtered Pack of 960
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Product Code. 12736769 Supplier Axygen™ Supplier No. ZTF100RS
  • ONLINE EXCLUSIVE PRICE
    810.00€
    661.02€ / Pack of 4800
    Save 148.98€
    18% Off
Estimated Shipment: 08-12-2026
Add to basket
Add to basket
Bulk Quote Request
missing translation for 'validMainframeMsg'
Clear, autoclavable tips; color-coded racks.
  • Tip format: 96 tip and 384 tip
  • Tip volume range: 25 μL to 200 μL
  • Tip material: Clear polypropylene
  • RNase-/DNase-/human gDNA-free
  • Nonpyrogenic
  • Available options:
    - With or without aerosol-resistant filters
    - Nonsterile or sterile
    - Standard or Axygen™ Maxymum Recovery™ surface
  • Compatible heads: HV96, CAS-4200-1 (20 - 150 μL/sec.), LV96, LV384
  • Rack format: Non-ANSI (96) and ANSI/SLAS (384)
  • Rack material: Polypropylene
Order Info
96 Tips/Rack, 10 Racks/Pack, 5 Packs/Case
TRUSTED_SUSTAINABILITY
Specifications
Volume (Metric) 100 μL
Filtered Filtered
Sterility Sterile
Color Clear
Rack Color Gray
Autoclavable Autoclavable
Certifications/Compliance Non-ANSI
Format Rack
Tip Style Standard, Round
Packaging Non-SBS Rack
Quantity 50 Racks/Cs.
For Use With (Equipment) Tecan Freedom EVO, Tecan TeMo, Tecan Aquarius MCA96 head, Zymark/Caliper RapidPlate, SciClone, SciClone HV96, CAS-4200-1ch. 20-150μL/sec. head
Product Line Caliper Zymark
Material Polyethylene