CAS RN 1037184-07-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
(2RS)-2,6-Dichloro-N-(2,6-dimethylphenyl)hexanamide, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
2,6-Dichloro-N-(2,6-dimethylphenyl)hexanamide, TRC
CAS: 1037184-07-8 Molecular Formula: C14 H19 Cl2 N O Molecular Weight (g/mol): 288.21 Synonym: (2RS)-2,6-Dichloro-N-(2,6-dimethylphenyl)hexanamide,Bupivacaine Hydrochloride Imp. D (EP) IUPAC Name: 2,6-dichloro-N-(2,6-dimethylphenyl)hexanamide SMILES: Cc1cccc(C)c1NC(=O)C(Cl)CCCCCl