missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichloro-N-(2,6-dimethylphenyl)hexanamide, TRC

Product Code. 30137885
Change view
Click to view available options
Quantity:
25 mg
250 mg
Unit Size:
250mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137885 250 mg 250mg
30011270 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30137885 Supplier Toronto Research Chemicals Supplier No. TRCD434225250MG
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1037184-07-8
Molecular Formula C14 H19 Cl2 N O
Molecular Weight (g/mol) 288.21
Synonym (2RS)-2,6-Dichloro-N-(2,6-dimethylphenyl)hexanamide, Bupivacaine Hydrochloride Imp. D (EP)
IUPAC Name 2,6-dichloro-N-(2,6-dimethylphenyl)hexanamide
SMILES Cc1cccc(C)c1NC(=O)C(Cl)CCCCCl

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C14H19Cl2NO/c1-10-6-5-7-11(2)13(10)17-14(18)12(16)8-3-4-9-15/h5-7,12H,3-4,8-9H2,1-2H3,(H,17,18)
Formula Weight 287.0844
Percent Purity >95%
Chemical Name or Material (2RS)-2,6-Dichloro-N-(2,6-dimethylphenyl)hexanamide
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.