accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 112-15-2

IUPAC Name: 2-(2-ethoxyethoxy)ethyl acetate
Synonyms: carbitol acetate ethoxydiglycol acetate
Molecular Weight (g/mol): 176.21
Molecular Formula: C8H16O4
InChi Key: FPZWZCWUIYYYBU-UHFFFAOYSA-N
SMILES: CCOCCOCCOC(C)=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (3)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (2)