missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Carbitol Acetate, TRC

Product Code. 30378749
Change view
Click to view available options
Quantity:
25 g
50 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30378749 25 g
30375724 50 g
30376233 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30378749 Supplier Toronto Research Chemicals Supplier No. TRCE49597525G
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-15-2
Molecular Formula C8 H16 O4
Molecular Weight (g/mol) 176.21
Synonym 2-(2-Ethoxyethoxy)ethyl Acetate, Carbitol acetate, Diethylene Glycol Ethyl Ether Acetate, Ektasolve DE acetate, NSC 8702NSC 870
IUPAC Name 2-(2-ethoxyethoxy)ethyl acetate
SMILES CCOCCOCCOC(=O)C

Specifications

Quantity 25 g
InChI Formula InChI=1S/C8H16O4/c1-3-10-4-5-11-6-7-12-8(2)9/h3-7H2,1-2H3
Formula Weight 176.1049
Chemical Name or Material Ethyl Carbitol Acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.