CAS RN 1141-23-7
IUPAC Name:
4-carbamoyl-3-(4-chlorophenyl)butanoic acid
Synonyms:
4-carbamoyl-3-(4-chlorophenyl)butanoic acid
Molecular Weight (g/mol):
241.67
Molecular Formula:
C11H12ClNO3
InChi Key:
NLCJVRPOYTZTMS-UHFFFAOYNA-N
SMILES:
NC(=O)CC(CC(O)=O)C1=CC=C(Cl)C=C1
1
–
2
of
2
results
1
(3RS)-5-Amino-3-(4-chlorophenyl)-5-oxopentanoic Acid, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
3-(4-Chlorophenyl)glutaramic Acid, TRC
CAS: 1141-23-7 Molecular Formula: C11 H12 Cl N O3 Molecular Weight (g/mol): 241.67 Synonym: (3RS)-5-Amino-3-(4-chlorophenyl)-5-oxopentanoic Acid,Baclofen Imp. B (EP) IUPAC Name: 5-amino-3-(4-chlorophenyl)-5-oxopentanoic acid SMILES: NC(=O)CC(CC(=O)O)c1ccc(Cl)cc1