missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Chlorophenyl)glutaramic Acid, TRC

Product Code. 30137805
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137805 5 g 5g
30136419 10 g 10g
30012663 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137805 Supplier Toronto Research Chemicals Supplier No. TRCC3781305G
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1141-23-7
Molecular Formula C11 H12 Cl N O3
Molecular Weight (g/mol) 241.67
Synonym (3RS)-5-Amino-3-(4-chlorophenyl)-5-oxopentanoic Acid, Baclofen Imp. B (EP)
IUPAC Name 5-amino-3-(4-chlorophenyl)-5-oxopentanoic acid
SMILES NC(=O)CC(CC(=O)O)c1ccc(Cl)cc1

Specifications

Quantity 5 g
InChI Formula InChI=1S/C11H12ClNO3/c12-9-3-1-7(2-4-9)8(5-10(13)14)6-11(15)16/h1-4,8H,5-6H2,(H2,13,14)(H,15,16)
Formula Weight 241.0506
Chemical Name or Material (3RS)-5-Amino-3-(4-chlorophenyl)-5-oxopentanoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.