accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 115-77-5

IUPAC Name: 2,2-bis(hydroxymethyl)propane-1,3-diol
Synonyms: pentek pentaerythritol
Molecular Weight (g/mol): 136.15
Molecular Formula: C5H12O4
InChi Key: WXZMFSXDPGVJKK-UHFFFAOYSA-N
SMILES: OCC(CO)(CO)CO
Molecular Weight (g/mol) 
  • (3)
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)