CAS RN 1173020-63-7
CAS RN 1173020-63-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Guanidineacetic-2,2-d2 Acid, CDN
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
2
Guanidineacetic-2,2-d2 Acid, TRC
CAS: 1173020-63-7 Molecular Formula: C3 D2 H5 N3 O2 Molecular Weight (g/mol): 119.1189 Synonym: Glycine-2,2-d2, N-(aminoiminomethyl)- (ACI),N-(Aminoiminomethyl)glycine-2,2-d2 (ACI) IUPAC Name: 2-carbamimidamido-2,2-dideuterioacetic acid SMILES: [2H]C([2H])(NC(=N)N)C(=O)O