missing translation for 'onlineSavingsMsg'
Learn More

Guanidineacetic-2,2-d2 Acid, TRC

Product Code. 30240940
Change view
Click to view available options
Quantity:
2.5 mg
5 mg
25 mg
Unit Size:
2.5mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30240940 2.5 mg 2.5mg
30240929 25 mg 25mg
30240844 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30240940 Supplier Toronto Research Chemicals Supplier No. TRCG8212512.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1173020-63-7
Molecular Formula C3 D2 H5 N3 O2
Molecular Weight (g/mol) 119.1189
Synonym Glycine-2,2-d2, N-(aminoiminomethyl)- (ACI), N-(Aminoiminomethyl)glycine-2,2-d2 (ACI)
IUPAC Name 2-carbamimidamido-2,2-dideuterioacetic acid
SMILES [2H]C([2H])(NC(=N)N)C(=O)O

Specifications

Quantity 2.5 mg
InChI Formula InChI=1S/C3H7N3O2/c4-3(5)6-1-2(7)8/h1H2,(H,7,8)(H4,4,5,6)/i1D2
Formula Weight 119.0664
Chemical Name or Material Guanidineacetic-2,2-d2 Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.