accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 118-79-6

IUPAC Name: 2,4,6-tribromophenol
Synonyms: 2,4,6-tribromophenol flammex 3BP bromkal pur 3 tribromophenol bromol C6H3br3O
Molecular Weight (g/mol): 330.8
Molecular Formula: C6H3Br3O
InChi Key: BSWWXRFVMJHFBN-UHFFFAOYSA-N
SMILES: OC1=C(Br)C=C(Br)C=C1Br
Molecular Weight (g/mol) 
  • (3)
  • (16)
Solvent or Matrix 
  • (1)
  • (2)
  • (4)
  • (2)
  • (8)
  • (1)
Concentration 
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
Certifications/Compliance 
  • (3)
  • (5)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (9)
  • (9)
  • (3)
  • (4)
  • (3)
  • (2)
  • (3)
  • (16)
  • (1)
  • (2)
  • (4)
  • (2)
  • (8)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (3)
  • (5)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (3)
  • (2)