accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH3FeOHOOOOOO

CAS RN 1185-57-5

IUPAC Name: iron(3+) 2-hydroxypropane-1,2,3-tricarboxylate amine
Synonyms: ferric ammonium citrate 枸橼酸铁铵
Molecular Weight (g/mol): 261.98
Molecular Formula: C6H8FeNO7
InChi Key: FRHBOQMZUOWXQL-UHFFFAOYSA-K
SMILES: N.[Fe+3].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O
Molecular Weight (g/mol) 
  • (4)
  • (10)
  • (4)
Quantity 
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (5)
  • (1)
Physical Form 
  • (9)
  • (1)
  • (2)
Percent Purity 
  • (4)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (2)
  • (4)
  • (3)
  • (2)
  • (2)
  • (4)
  • (10)
  • (4)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (5)
  • (1)
  • (9)
  • (1)
  • (2)
  • (4)
  • (3)