accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COOOCH3

CAS RN 120-44-5

IUPAC Name: 1,2-bis(4-methoxyphenyl)ethan-1-one
Synonyms: 1,2-bis(4-methoxyphenyl)ethanone
Molecular Weight (g/mol): 256.3
Molecular Formula: C16H16O3
InChi Key: SICBLYCPRWNHHP-UHFFFAOYSA-N
SMILES: COC1=CC=C(CC(=O)C2=CC=C(OC)C=C2)C=C1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results