accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1222-05-5

IUPAC Name: 4,6,6,7,8,8-hexamethyl-1H,3H,4H,6H,7H,8H-indeno[5,6-c]pyran
Synonyms: galaxolide hexamethylindanopyran
Molecular Weight (g/mol): 258.41
Molecular Formula: C18H26O
InChi Key: ONKNPOPIGWHAQC-UHFFFAOYNA-N
SMILES: CC1C(C)(C)C2=C(C=C3C(C)COCC3=C2)C1(C)C
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results