missing translation for 'onlineSavingsMsg'
Learn More

Galaxolide (Mixture of Diastereomers), TRC

Product Code. 17970485
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17970485 100 mg 100mg
17940485 50 mg 50mg
17941415 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17970485 Supplier Toronto Research Chemicals Supplier No. TRCG189000100MG
 Restricted Item
Estimated Shipment: 28-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1222-05-5
Molecular Formula C18 H26 O
Molecular Weight (g/mol) 258.4
Synonym 1,3,4,6,7,8-Hexahydro-4,6,6,7,8,8-hexamethylcyclopenta[g]-2-benzopyran, 1,3,4,6,7,8-Hexahydro-4,6,6,7,8,8-hexamethylcyclopenta[g]-2-benzopyrane, 1,3,4,6,7,8-Hexahydro-4,6,6,8,8,8-hexamethylcyclopenta-2-benzopyran, 4,6,6,7,8,8-Hexamethyl-1,3,4,6,7,8-hexahydrocyclopenta[g]isochromene, Abbalide, Galaxolide, Galaxolide 50, Galaxolide 50BB, Galaxolide 50IPM, Galaxolide White, HHCB, Pearlide, Cyclopenta[g]-2-benzopyran, 1,3,4,6,7,8-hexahydro-4,6,6,7,8,8-hexamethyl-
IUPAC Name 4,6,6,7,8,8-hexamethyl-1,3,4,7-tetrahydrocyclopenta[g]isochromene
SMILES CC1COCc2cc3c(cc12)C(C)(C)C(C)C3(C)C

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C18H26O/c1-11-9-19-10-13-7-15-16(8-14(11)13)18(5,6)12(2)17(15,3)4/h7-8,11-12H,9-10H2,1-6H3
Formula Weight 258.1984
Chemical Name or Material HHCB (Galaxolide)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.