accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3O

CAS RN 123-72-8

IUPAC Name: butanal
Synonyms: butoxide fema number 2219 butalyde butyraldehyde (substance) butyraldehyd n-C3H7cho aldehyde butyrique butyrylaldehyde aldeide butirrica butylaldehyde
Molecular Weight (g/mol): 72.11
Molecular Formula: C4H8O
InChi Key: ZTQSAGDEMFDKMZ-UHFFFAOYSA-N
SMILES: CCCC=O
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (3)