accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H2OH2OH2OH2OH2OH2OH2OH2OH2OH2ONaNaNaNaOPOOOPOOO

CAS RN 13472-36-1

IUPAC Name: tetrasodium decahydrate (phosphonatooxy)phosphonate
Synonyms: tetrasodium pyrophosphate decahydrate
Molecular Weight (g/mol): 446.05
Molecular Formula: H20Na4O17P2
InChi Key: VZWGHDYJGOMEKT-UHFFFAOYSA-J
SMILES: O.O.O.O.O.O.O.O.O.O.[Na+].[Na+].[Na+].[Na+].[O-]P([O-])(=O)OP([O-])([O-])=O
Molecular Weight (g/mol) 
  • (22)
  • (2)
Quantity 
  • (1)
  • (1)
  • (4)
  • (1)
  • (1)
  • (4)
  • (1)
  • (2)
  • (1)
  • (6)
  • (1)
  • (1)
Grade 
  • (3)
  • (5)
  • (4)
  • (1)
  • (1)
  • (4)
Percent Purity 
  • (3)
  • (9)
  • (1)
  • (3)
Physical Form 
  • (3)
  • (5)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (4)
  • (2)
  • (3)
  • (22)
  • (2)
  • (1)
  • (1)
  • (4)
  • (1)
  • (1)
  • (4)
  • (1)
  • (2)
  • (1)
  • (6)
  • (1)
  • (1)
  • (3)
  • (5)
  • (4)
  • (1)
  • (1)
  • (4)
  • (3)
  • (9)
  • (1)
  • (3)
  • (3)
  • (5)
  • (2)