accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CONNNHNN(S)CH3H3COHO

CAS RN 137862-53-4

IUPAC Name: (2S)-3-methyl-2-(N-{[2'-(2H-1,2,3,4-tetrazol-5-yl)-[1,1'-biphenyl]-4-yl]methyl}pentanamido)butanoic acid
Synonyms: valsartan walsartan tareg cepa brand of valsartan Vals kalpress schwarz brand of valsartan valsartan (product) miten provas
Molecular Weight (g/mol): 435.53
Molecular Formula: C24H29N5O3
InChi Key: ACWBQPMHZXGDFX-QFIPXVFZSA-N
SMILES: CCCCC(=O)N(CC1=CC=C(C=C1)C1=CC=CC=C1C1=NNN=N1)[C@@H](C(C)C)C(O)=O
Molecular Weight (g/mol) 
  • (4)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Certifications/Compliance 
  • (1)
  • (1)
Industry Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (2)
  • (4)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)