missing translation for 'onlineSavingsMsg'
Learn More

Valsartan, TRC

Product Code. 30041193
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30041193 10 mg 10mg
30040621 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30041193 Supplier Toronto Research Chemicals Supplier No. TRCV09575010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 137862-53-4
Molecular Formula C24 H29 N5 O3
Molecular Weight (g/mol) 435.52
Synonym L-Valine, N-(1-oxopentyl)-N-[[2'-(1H-tetrazol-5-yl)[1,1'-biphenyl]-4-yl]methyl]- (9CI), N-(1-Oxopentyl)-N-[[2'-(2H-tetrazol-5-yl)[1,1'-biphenyl]-4-yl]methyl]-L-valine, CGP 48933, Cardopan, Diovan, Nisis, Tareg, Valsarect, Valsartan, Valsartanin
IUPAC Name (2S)-3-methyl-2-[pentanoyl-[[4-[2-(1H-tetrazol-5-yl)phenyl]phenyl]methyl]amino]butanoic acid
SMILES CCCCC(=O)N(Cc1ccc(cc1)c2ccccc2c3nnn[nH]3)[C@@H](C(C)C)C(=O)O

Specifications

InChI Formula InChI=1S/C24H29N5O3/c1-4-5-10-21(30)29(22(16(2)3)24(31)32)15-17-11-13-18(14-12-17)19-8-6-7-9-20(19)23-25-27-28-26-23/h6-9,11-14,16,22H,4-5,10,15H2,1-3H3,(H,31,32)(H,25,26,27,28)/t22-/m0/s1
Quantity 10 mg
Formula Weight 435.227
Percent Purity >95%
Shipping Condition Room Temperature
Chemical Name or Material Valsartan
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.