CAS RN 14040-05-2
IUPAC Name:
bis(5-hydroxy-2,2,6,6-tetramethylhept-4-en-3-one) copper
Synonyms:
bis(5-hydroxy-2,2,6,6-tetramethylhept-4-en-3-one) copper
Molecular Weight (g/mol):
432.1
Molecular Formula:
C22H40CuO4
InChi Key:
PEMCLTTUCGSLGJ-UHFFFAOYSA-N
SMILES:
[Cu].CC(C)(C)C(O)=CC(=O)C(C)(C)C.CC(C)(C)C(O)=CC(=O)C(C)(C)C
1
–
2
of
2
results
2
Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)copper(II), 99.9% (metals basis)
CAS: 14040-05-2 Molecular Formula: C22H38CuO4 Molecular Weight (g/mol): 430.09 MDL Number: MFCD00058920 InChI Key: JGMGCTZGOAONPQ-UHFFFAOYSA-N Synonym: copper bis 2,2,6,6-tetramethyl-3,5-heptanedionate,2,2,6,6-tetramethyl-5-2,2,6,6-tetramethyl-5-oxohept-3-en-3-yl oxy cuprio oxy hept-4-en-3-one PubChem CID: 91663146 SMILES: [Cu++].CC(C)(C)C(=O)[CH-]C(=O)C(C)(C)C.CC(C)(C)C(=O)[CH-]C(=O)C(C)(C)C