missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)copper(II), 99%

Product Code. 11342468
Change view
Click to view available options
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
11342468 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11342468 Supplier Alfa Aesar™ Supplier No. 014516.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Vacuum deposition

TRUSTED_SUSTAINABILITY

Specifications

CAS 14040-05-2
Melting Point 198°C
Boiling Point 100°C (0.1 mmHg)(sublimation)
Molecular Formula C22H38CuO4
MDL Number MFCD00058920
Quantity 5 g
Synonym copper bis 2,2,6,6-tetramethyl-3,5-heptanedionate, 2,2,6,6-tetramethyl-5-2,2,6,6-tetramethyl-5-oxohept-3-en-3-yl oxy cuprio oxy hept-4-en-3-one
InChI Key JGMGCTZGOAONPQ-UHFFFAOYSA-N
SMILES [Cu++].CC(C)(C)C(=O)[CH-]C(=O)C(C)(C)C.CC(C)(C)C(=O)[CH-]C(=O)C(C)(C)C
IUPAC Name copper(2+) bis(2,2,6,6-tetramethyl-3,5-dioxoheptan-4-ide)
Molecular Weight (g/mol) 430.09
PubChem CID 91663146
Formula Weight 430.05
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)copper(II)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.