CAS RN 141-73-1
CAS RN 141-73-1
IUPAC Name:
4-methoxy-4-methylpentan-2-ol
Synonyms:
4-methoxy-4-methylpentan-2-ol
Molecular Weight (g/mol):
132.2
Molecular Formula:
C7H16O2
InChi Key:
OMSLSFZIEVFEIH-UHFFFAOYNA-N
SMILES:
COC(C)(C)CC(C)O
1
–
1
of
1
results
1
4-Methoxy-4-methyl-2-pentanol, 98%, Thermo Scientific™
CAS: 141-73-1 Molecular Formula: C7H16O2 Molecular Weight (g/mol): 132.20 MDL Number: MFCD00046654 InChI Key: OMSLSFZIEVFEIH-UHFFFAOYNA-N PubChem CID: 78955 IUPAC Name: 4-methoxy-4-methylpentan-2-ol SMILES: COC(C)(C)CC(C)O