missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxy-4-methyl-2-pentanol, 98%, Thermo Scientific™

Product Code. 11496067
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11496067 5 g 5g
11406077 25 g 25g
11416077 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11496067 Supplier Thermo Scientific Alfa Aesar Supplier No. B25078.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 141-73-1
Molecular Formula C7H16O2
Molecular Weight (g/mol) 132.20
MDL Number MFCD00046654
InChI Key OMSLSFZIEVFEIH-UHFFFAOYNA-N
PubChem CID 78955
IUPAC Name 4-methoxy-4-methylpentan-2-ol
SMILES COC(C)(C)CC(C)O

Specifications

Quantity 5 g
Synonym 4-methoxy-4-methyl-2-pentanol, pent-oxol solvent, pent-oxol, 2-pentanol, 4-methoxy-4-methyl, 4-01-00-02566 beilstein handbook reference, 4-methoxy-4-methyl-pentan-2-ol, 2-pentanol,4-methoxy-4-methyl
Formula Weight 132.2
Percent Purity 98%
Chemical Name or Material 4-Methoxy-4-methyl-2-pentanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.