CAS RN 143130-82-9
IUPAC Name:
methyl 4-{2-[4-(methoxycarbonyl)phenyl]ethenyl}benzoate
Synonyms:
methyl 4-{2-[4-(methoxycarbonyl)phenyl]ethenyl}benzoate
Molecular Weight (g/mol):
296.32
Molecular Formula:
C18H16O4
InChi Key:
JOODVYOWCWQPMV-UHFFFAOYSA-N
SMILES:
COC(=O)C1=CC=C(C=CC2=CC=C(C=C2)C(=O)OC)C=C1
1
–
1
of
1
results
1
Dimethyl cis-stilbene-4,4'-dicarboxylate
CAS: 143130-82-9 Molecular Formula: C18H16O4 Molecular Weight (g/mol): 296.322 MDL Number: MFCD00082669 InChI Key: JOODVYOWCWQPMV-ARJAWSKDSA-N Synonym: dimethyl cis-stilbene-4,4'-dicarboxylate,dimethyl cis-stilbene-4,4/'-dicarboxylate,z-dimethyl 4,4'-ethene-1,2-diyl dibenzoate,z-4,4'-stilbenedicarboxylic acid dimethyl ester,methyl 4-z-2-4-methoxycarbonylphenyl ethenyl benzoate,methyl 4-z-2-4-methoxycarbonyl phenyl ethenyl benzoate PubChem CID: 7556518 IUPAC Name: methyl 4-[(Z)-2-(4-methoxycarbonylphenyl)ethenyl]benzoate SMILES: COC(=O)C1=CC=C(C=C1)C=CC2=CC=C(C=C2)C(=O)OC