accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OHNOCH3

CAS RN 14430-23-0

IUPAC Name: 5,6-dimethoxy-1H-indole
Synonyms: 5,6-dimethoxy-1H-indole
Molecular Weight (g/mol): 177.2
Molecular Formula: C10H11NO2
InChi Key: QODBZRNBPUPLEZ-UHFFFAOYSA-N
SMILES: COC1=C(OC)C=C2C=CNC2=C1
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)