missing translation for 'onlineSavingsMsg'
Learn More

5,6-Dimethoxyindole, 98%, Thermo Scientific Chemicals

Product Code. 11474607
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11474607 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11474607 Supplier Thermo Scientific Alfa Aesar Supplier No. B21126.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

It is applied as a reactant in synthesis of indolylhydroxyoxindoles via enantioselective Friedel-Crafts reaction, in synthesis of benzyl trimethoxyindoles, for synthesis of benzoylpiperazinyl-indolyl ethane dione derivatives as HIV-1 inhibitors, for synthesis of 1-aroylindole 3-aroylindoles combretastatin A-4 analogs as antitumor agents and tubulin polymerization inhibitors and for preparation of tryptophanol derivatives via the Grignard reaction. It is an indole derivative as analgesic anti-inflammatory useful in treatment of pain, inflammation. It is also used in the synthesis of N-benzyl-N-cyclopropyl-5,6-dimethoxyindole-3-glyoxalamide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is applied as a reactant in synthesis of indolylhydroxyoxindoles via enantioselective Friedel-Crafts reaction, in synthesis of benzyl trimethoxyindoles, for synthesis of benzoylpiperazinyl-indolyl ethane dione derivatives as HIV-1 inhibitors, for synthesis of 1-aroylindole 3-aroylindoles combretastatin A-4 analogs as antitumor agents and tubulin polymerization inhibitors and for preparation of tryptophanol derivatives via the Grignard reaction. It is an indole derivative as analgesic anti-inflammatory useful in treatment of pain, inflammation. It is also used in the synthesis of N-benzyl-N-cyclopropyl-5,6-dimethoxyindole-3-glyoxalamide.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14430-23-0
Molecular Formula C10H11NO2
Molecular Weight (g/mol) 177.20
MDL Number MFCD00005675
InChI Key QODBZRNBPUPLEZ-UHFFFAOYSA-N
PubChem CID 84431
IUPAC Name 5,6-dimethoxy-1H-indole
SMILES COC1=C(OC)C=C2C=CNC2=C1

Specifications

Melting Point 154°C to 160°C
Boiling Point 198°C (8 mmHg)
Quantity 1 g
Beilstein 5683
Synonym 5,6-dimethoxyindole, 1h-indole, 5,6-dimethoxy, 5,6-dimethoxyindol, zlchem 178, pubchem7329, acmc-1cbhq, 1h-indole,5,6-dimethoxy, #
Solubility Information Insoluble in water.
Formula Weight 177.2
Percent Purity 98%
Chemical Name or Material 5,6-Dimethoxyindole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.